COMH42123F28 Combi-Blocks, Inc.

6-Chloro-2-(chloromethyl)imidazo[1,2-a]pyridine, HCl

  • MDL number MFCD07841696
  • Cas Number : 502841-53-4
  • Molecular Weight : 237.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2308
Availability details for
Quantity
  • Availability
  • Add to Cart