COMH42123F3B Combi-Blocks, Inc.

5-(1H-Benzoimidazol-2-yl)-2-methyl-phenylamine

  • MDL number MFCD00852063
  • Cas Number : 292644-33-8
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3525

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3525
Availability details for
Quantity
  • Availability
  • Add to Cart