COMH42123F3E Combi-Blocks, Inc.

4-Azepan-1-ylaniline, HCl

  • MDL number MFCD06683755
  • Cas Number : 57356-18-0
  • Molecular Weight : 226.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3771
Availability details for
Quantity
  • Availability
  • Add to Cart