COMH42123F47 Combi-Blocks, Inc.

Sodium 3-amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD08691308
  • Cas Number : 144254-93-3
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4110
Availability details for
Quantity
  • Availability
  • Add to Cart