COMH42123F4E Combi-Blocks, Inc.

4-(Pyrrolidin-2-ylcarbonyl)morpholine

  • MDL number MFCD06740216
  • Cas Number : 1096446-01-3
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4334

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4334
Availability details for
Quantity
  • Availability
  • Add to Cart