COMH42123F58 Combi-Blocks, Inc.

Methyl 2-bromo-5-sulfamoylbenzoate

  • MDL number MFCD11057846
  • Cas Number : 924867-88-9
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4713
Availability details for
Quantity
  • Availability
  • Add to Cart