COMH42123F66 Combi-Blocks, Inc.

5-tert-Butyl-1h-imidazole

  • MDL number MFCD18711525
  • Cas Number : 21149-98-4
  • Molecular Weight : 124.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5248
Availability details for
Quantity
  • Availability
  • Add to Cart