COMH42123F78 Combi-Blocks, Inc.

3-(2-Methoxyphenoxy)propylamine

  • MDL number MFCD05053657
  • Cas Number : 3245-88-3
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6165
Availability details for
Quantity
  • Availability
  • Add to Cart