COMH42123F88 Combi-Blocks, Inc.

3-Pyrrolidin-1-yl-benzoic acid methyl ester

  • MDL number MFCD06740075
  • Cas Number : 186086-71-5
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6881
Availability details for
Quantity
  • Availability
  • Add to Cart