COMH42123FA3 Combi-Blocks, Inc.

N-Cycloheptyl-N-methylamine

  • MDL number MFCD08271751
  • Cas Number : 42870-65-5
  • Molecular Weight : 127.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8462
Availability details for
Quantity
  • Availability
  • Add to Cart