COMH42123FB2 Combi-Blocks, Inc.

4-(4-tert-Butylphenoxymethyl)benzoic acid

  • MDL number MFCD02090849
  • Cas Number : 132991-24-3
  • Molecular Weight : 284.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9380
Availability details for
Quantity
  • Availability
  • Add to Cart