COMH42123FBE Combi-Blocks, Inc.

2-[(Cyclopropylcarbonyl)amino]benzoic acid

  • MDL number MFCD02602218
  • Cas Number : 40057-21-4
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9650
Availability details for
Quantity
  • Availability
  • Add to Cart