COMH42123FC1 Combi-Blocks, Inc.

1-(1H-Benzimidazol-2-yl)-2-methylpropan-1-amine, HCl

  • MDL number MFCD08445277
  • Cas Number : 1177315-00-2
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9752
Availability details for
Quantity
  • Availability
  • Add to Cart