COMH42123FC7 Combi-Blocks, Inc.

4-Methoxythiophene-2-carbaldehyde

  • MDL number MFCD24052836
  • Cas Number : 98316-32-6
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7336
Availability details for
Quantity
  • Availability
  • Add to Cart