COMH42123FC9 Combi-Blocks, Inc.

2-(N,N-Dimethylamino)-3-fluoropyridine

  • MDL number MFCD20327984
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WA-6069

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WA-6069
Availability details for
Quantity
  • Availability
  • Add to Cart