COMH42123FCF Combi-Blocks, Inc.

7-Nitro-2,4-dihydroisoquinoline-1,3-dione

  • MDL number MFCD11227005
  • Cas Number : 611187-01-0
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9690
Availability details for
Quantity
  • Availability
  • Add to Cart