COMH580D80C5 Combi-Blocks, Inc.

2-(dihydroxyboranyl)-5-(trifluoromethyl)benzoic acid

  • Molecular Weight : 233.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4839
Availability details for
Quantity
  • Availability
  • Add to Cart