COMH580D80C7 Combi-Blocks, Inc.

[4-(1-Methylpiperidin-4-yl)phenyl]boronic acid

  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1419
Availability details for
Quantity
  • Availability
  • Add to Cart