COMH580D80C8 Combi-Blocks, Inc.

(3-{[(tert-Butoxy)carbonyl]amino}-5-cyanophenyl)boronic acid

  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4556
Availability details for
Quantity
  • Availability
  • Add to Cart