COMH580D80C9 Combi-Blocks, Inc.

1-(4-Methylpiperazin-1-yl)eth-1-ene-1,2,2-tricarbonitrile

  • Cas Number : 2377920-17-5
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4810
Availability details for
Quantity
  • Availability
  • Add to Cart