COMH580D80CD Combi-Blocks, Inc.

2-(4-Iodophenyl)acetamide

  • Cas Number : 84863-81-0
  • Molecular Weight : 261.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4830
Availability details for
Quantity
  • Availability
  • Add to Cart