COMH580D80CF Combi-Blocks, Inc.

2-(4-Amino-2-bromophenoxy)-N-cyclopropylacetamide

  • Cas Number : 2378806-55-2
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-amino-2-bromophenoxy)-N-cyclopropylacetamide

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4872

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4872
Availability details for
Quantity
  • Availability
  • Add to Cart