COMH580D80D1 Combi-Blocks, Inc.

2-(4-Amino-2-bromophenoxy)-N,N-dimethylacetamide

  • Cas Number : 2378806-59-6
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-amino-2-bromophenoxy)-N,N-dimethylacetamide

Description

Purity 96%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4878

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4878
Availability details for
Quantity
  • Availability
  • Add to Cart