COMH580D80D2 Combi-Blocks, Inc.

1-[(4-Methoxyphenyl)methyl]-3-methylpyrrolidin-2-one

  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7464
Availability details for
Quantity
  • Availability
  • Add to Cart