COMH580D80DF Combi-Blocks, Inc.

(2-Cyanopyridin-4-yl)boronic acid

  • MDL number MFCD10696640
  • Cas Number : 903513-60-0
  • Molecular Weight : 147.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0143
Availability details for
Quantity
  • Availability
  • Add to Cart