COMH580D80E3 Combi-Blocks, Inc.

(2-fluoro-5-hydroxypyridin-3-yl)boronic acid

  • MDL number MFCD16611107
  • Cas Number : 2305898-10-4
  • Molecular Weight : 156.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6566
Availability details for
Quantity
  • Availability
  • Add to Cart