COMH580D80E4 Combi-Blocks, Inc.

[2-fluoro-6-(morpholin-4-yl)pyridin-3-yl]boronic acid

  • MDL number MFCD18261461
  • Cas Number : 955374-17-1
  • Molecular Weight : 226.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6739
Availability details for
Quantity
  • Availability
  • Add to Cart