COMH580D80F3 Combi-Blocks, Inc.

Methyl 2-[2-(2-methoxy-2-oxoethyl)phenyl]acetate

  • Cas Number : 19109-69-4
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6001
Availability details for
Quantity
  • Availability
  • Add to Cart