COMH580D80FD Combi-Blocks, Inc.

(5-Fluoro-2-methylpyridin-4-yl)boronic acid

  • MDL number MFCD07374988
  • Cas Number : 1310404-23-9
  • Molecular Weight : 154.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1393

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1393
Availability details for
Quantity
  • Availability
  • Add to Cart