COMH580D8100 Combi-Blocks, Inc.

[5-Methyl-2-(2,2,2-trifluoroethoxy)phenyl]boranediol

  • MDL number MFCD11183204
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1984
Availability details for
Quantity
  • Availability
  • Add to Cart