COMH580D8101 Combi-Blocks, Inc.

[5-Methyl-2-(3-methylbutoxy)phenyl]boranediol

  • MDL number MFCD11183205
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1985

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1985
Availability details for
Quantity
  • Availability
  • Add to Cart