COMH580D8102 Combi-Blocks, Inc.

[5-Methyl-2-(oxolan-2-ylmethoxy)phenyl]boranediol

  • MDL number MFCD11183207
  • Cas Number : 1311159-17-7
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2001
Availability details for
Quantity
  • Availability
  • Add to Cart