COMH580D8103 Combi-Blocks, Inc.

[2-(2-Methoxyethoxy)-5-methylphenyl]boranediol

  • MDL number MFCD11183208
  • Cas Number : 1311185-50-8
  • Molecular Weight : 210.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2002
Availability details for
Quantity
  • Availability
  • Add to Cart