COMH580D8104 Combi-Blocks, Inc.

[6-(Pyrazol-1-yl)pyridin-3-yl]boronic acid

  • MDL number MFCD18260602
  • Molecular Weight : 189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3495
Availability details for
Quantity
  • Availability
  • Add to Cart