COMH580D8105 Combi-Blocks, Inc.

5-(2-Thiazolyl)thiophene-2-boronic acid

  • MDL number MFCD12032294
  • Cas Number : 1872434-79-1
  • Molecular Weight : 211.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3862
Availability details for
Quantity
  • Availability
  • Add to Cart