COMH580D8106 Combi-Blocks, Inc.

2-(cyclopropylmethoxy)-5-methylphenylboronic acid

  • MDL number MFCD12138947
  • Molecular Weight : 206.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3917
Availability details for
Quantity
  • Availability
  • Add to Cart