COMH580D8107 Combi-Blocks, Inc.

(5-Methyl-2-[2-(morpholin-4-yl)ethoxy]phenyl)boranediol

  • MDL number MFCD12558056
  • Cas Number : 1311182-84-9
  • Molecular Weight : 265.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-2-[2-(4-morpholinyl)ethoxy]phenylboronic acid
Inchi Code 1S/C13H20BNO4/c1-11-2-3-13(12(10-11)14(16)17)19-9-6-15-4-7-18-8-5-15/h2-3,10,16-17H,4-9H2,1H3
Inchi Key KEECYKFFPMVRQY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4034
Availability details for
Quantity
  • Availability
  • Add to Cart