COMH580D8109 Combi-Blocks, Inc.

B-[6-(Dimethylamino)-2-fluoro-3-pyridinyl]-boronic acid

  • MDL number MFCD12964581
  • Cas Number : 909187-40-2
  • Molecular Weight : 184.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4269
Availability details for
Quantity
  • Availability
  • Add to Cart