COMH580D810B Combi-Blocks, Inc.

3-[(2,2,2-Trifluoroethoxy)methyl]phenylboronic acid

  • MDL number MFCD14687254
  • Cas Number : 1334218-47-1
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4617

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4617
Availability details for
Quantity
  • Availability
  • Add to Cart