COMH580D810C Combi-Blocks, Inc.

3,4-Bis(tert-butoxycarbonylamino)phenylboronic acid

  • MDL number MFCD18383584
  • Cas Number : 1233693-22-5
  • Molecular Weight : 352.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-bis[(tert-butoxycarbonyl)amino]phenylboronic acid
Inchi Code 1S/C16H25BN2O6/c1-15(2,3)24-13(20)18-11-8-7-10(17(22)23)9-12(11)19-14(21)25-16(4,5)6/h7-9,22-23H,1-6H3,(H,18,20)(H,19,21)
Inchi Key OLHUPXIXFDSXGK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-7412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-7412
Availability details for
Quantity
  • Availability
  • Add to Cart