COMH580D810F Combi-Blocks, Inc.

2-Methyl-5-(thiazol-2-yl)phenylboronic acid

  • MDL number MFCD18383563
  • Cas Number : 1235963-01-5
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2380
Availability details for
Quantity
  • Availability
  • Add to Cart