COMH580D8112 Combi-Blocks, Inc.

2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD18383388
  • Cas Number : 1258440-84-4
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-1089

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-1089
Availability details for
Quantity
  • Availability
  • Add to Cart