COMH580D8113 Combi-Blocks, Inc.

3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol

  • MDL number MFCD16995272
  • Cas Number : 2256755-09-4
  • Molecular Weight : 255.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-1767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-1767
Availability details for
Quantity
  • Availability
  • Add to Cart