COMH580D8115 Combi-Blocks, Inc.

N-Isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

  • MDL number MFCD16995224
  • Molecular Weight : 290.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinecarboxamide
Inchi Code 1S/C15H23BN2O3/c1-10(2)18-13(19)12-9-11(7-8-17-12)16-20-14(3,4)15(5,6)21-16/h7-10H,1-6H3,(H,18,19)
Inchi Key AMOUHFHDJBXUMM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6789
Availability details for
Quantity
  • Availability
  • Add to Cart