COMH580D8116 Combi-Blocks, Inc.

Pyrrolidin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)methanone

  • MDL number MFCD16996042
  • Molecular Weight : 302.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-pyrrolidinylcarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Inchi Code 1S/C16H23BN2O3/c1-15(2)16(3,4)22-17(21-15)12-7-8-18-13(11-12)14(20)19-9-5-6-10-19/h7-8,11H,5-6,9-10H2,1-4H3
Inchi Key YEDQEFMBADBTMH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7248
Availability details for
Quantity
  • Availability
  • Add to Cart