COMH580D8117 Combi-Blocks, Inc.

Morpholino(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)methanone

  • MDL number MFCD16995585
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7949

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7949
Availability details for
Quantity
  • Availability
  • Add to Cart