COMH580D811C Combi-Blocks, Inc.

N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

  • MDL number MFCD11878165
  • Cas Number : 1443037-13-5
  • Molecular Weight : 276.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3251

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3251
Availability details for
Quantity
  • Availability
  • Add to Cart