COMH580D811D Combi-Blocks, Inc.

N-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

  • MDL number MFCD11878166
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3252
Availability details for
Quantity
  • Availability
  • Add to Cart