COMH580D811F Combi-Blocks, Inc.

6-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzothiazol-2-ylamine

  • MDL number MFCD13190219
  • Cas Number : 1244041-62-0
  • Molecular Weight : 276.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7075
Availability details for
Quantity
  • Availability
  • Add to Cart