COMH580D8120 Combi-Blocks, Inc.

[2-tert-Butoxycarbonylamino-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-carbamic acid tert-butyl ester

  • MDL number MFCD15527090
  • Molecular Weight : 434.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7412
Availability details for
Quantity
  • Availability
  • Add to Cart